Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Periodate ACS AR, Macron™

Catalog No. MK1129500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-1129-500 500 g Glass
MK-1129-125 125 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-1129-500 Supplier Avantor Supplier No. 112904

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 229.999
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
PubChem CID 516896
IUPAC Name potassium;periodate
SMILES [O-]I(=O)(=O)=O.[K+]

Specifications

Quantity 500 g
Formula Weight 230
Grade ACS
Packaging Glass
Chemical Name or Material Potassium Periodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.