accessibility menu, dialog, popup

UserName

Viewing profile as user:
KKOTeOO

CAS RN 7790-58-1

IUPAC Name: dipotassium tellurite
Synonyms: dipotassium tellurite
Molecular Weight (g/mol): 253.79
Molecular Formula: K2O3Te
InChi Key: BFPJYWDBBLZXOM-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-][Te]([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (4)
Grade 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (4)
  • (8)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)