Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium tellurite

Catalog No. 503126108 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6108 100 g
50-312-6020 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-6108 Supplier Sigma Aldrich Supplier No. P0677100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium tellurite is a hygroscopic granular white powder. It decomposes at 460-470oC.Potassium tellurite along with agar may be used as a component of a selective medium for growth of bacteria.

Specifications

CAS 7790-58-1
Quantity 100 g
Molecular Formula K2TeO3
Linear Formula K2TeO3
Synonym Dipotassium tellurite; Potassium tellurate; Potassium tellurium oxide
Molecular Weight (g/mol) 253.79
Percent Purity ≥90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.