CAS RN 7790-69-4
Lithium nitrate, anhydrous, 99%
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N Synonym: lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure PubChem CID: 10129889 ChEBI: CHEBI:63315 SMILES: [Li+].[O-][N+]([O-])=O
Lithium nitrate, 99+%, extra pure
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N Synonym: lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure PubChem CID: 10129889 ChEBI: CHEBI:63315 SMILES: [Li+].[O-][N+]([O-])=O
VeriSpec™ Lithium - Li+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N PubChem CID: 10129889 ChEBI: CHEBI:63315 IUPAC Name: lithium(1+) nitrate SMILES: [Li+].[O-][N+]([O-])=O
Lithium Nitrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N Synonym: lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure PubChem CID: 10129889 ChEBI: CHEBI:63315 IUPAC Name: lithium(1+) nitrate SMILES: [Li+].[O-][N+]([O-])=O
Lithium nitrate, Ionization Buffer Solution, Specpure™
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N PubChem CID: 10129889 ChEBI: CHEBI:63315 IUPAC Name: lithium;nitrate SMILES: [Li+].[O-][N+]([O-])=O
Lithium nitrate, 99.999%, (trace metal basis)
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N Synonym: lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure PubChem CID: 10129889 ChEBI: CHEBI:63315 SMILES: [Li+].[O-][N+]([O-])=O
Lithium nitrate hydrate, Puratronic™, 99.999% (metals basis)
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N Synonym: lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure PubChem CID: 10129889 ChEBI: CHEBI:63315 SMILES: [Li+].[O-][N+]([O-])=O
Lithium nitrate, anhydrous, 99.98% (metals basis)
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N Synonym: lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure PubChem CID: 10129889 ChEBI: CHEBI:63315 IUPAC Name: lithium;nitrate SMILES: [Li+].[O-][N+]([O-])=O
Lithium nitrate, anhydrous, 99.999% (metals basis)
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N Synonym: lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure PubChem CID: 10129889 ChEBI: CHEBI:63315 IUPAC Name: lithium;nitrate SMILES: [Li+].[O-][N+]([O-])=O
VeriSpec™ Lithium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N PubChem CID: 10129889 ChEBI: CHEBI:63315 IUPAC Name: lithium(1+) nitrate SMILES: [Li+].[O-][N+]([O-])=O
VeriSpec™ Mixed Cation Standard 9, Ricca Chemical
100 ppm Ba2+, Ca2+, K+, Li+, Mg2+, Mn2+, Na+, NH4+, Sr2+ /nManufactured and Tested in
VeriSpec™ Lithium (Li) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 7790-69-4 Molecular Formula: LiNO3 Molecular Weight (g/mol): 68.94 MDL Number: MFCD00011094 InChI Key: IIPYXGDZVMZOAP-UHFFFAOYSA-N PubChem CID: 10129889 ChEBI: CHEBI:63315 IUPAC Name: lithium(1+) nitrate SMILES: [Li+].[O-][N+]([O-])=O
- 1
- 2