Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Nitrate, Crystal, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611335 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-335 2.5 kg Amber Glass Bottle
18-611-330 25 g Amber Glass Bottle
18-611-331 125 g Amber Glass Bottle
18-611-333 500 g Amber Glass Bottle
18-611-332 12 kg Amber Glass Bottle
18-611-334 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-335 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L11202.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An effective oxidizing agent used in laboratory settings for research and in various technical applications.

  • Grade: Reagent
  • Molecular Formula: LiNO3
  • Assay (LiNO3): 99.0 %
  • Chloride (Cl): 0.02 %
  • Sulfates (SO4): 0.05 %
  • Water: 1.0 %
  • Deliquescent: Keep tightly closed
  • Physical State: Solid (Crystals, Granular)
  • Solubility: Soluble in water (15 g/L at 25°C)
  • CAS Number: 7790-69-4
  • Hazards: Oxidizer. Harmful if swallowed. Causes eye and skin irritation.
  • Storage: Keep tightly closed; store in a dry, well-ventilated area away from combustible and incompatible materials.
  • Applications: Used in fireworks and flares, laboratory research on crystal structures, and as an oxidizing agent in carbon dioxide absorption
  • Chemical Name: Lithium Nitrate
  • Formula: LiNO3
  • Appearance: Crystals, granular
  • Decomposition Temperature: 600 °C
  • Hazardous Properties: Oxidizing material; contact with combustible materials may cause fire
  • Regulatory Information: Manufactured under cGMP in U.S. facilities; ISO 9001:2015 certified
  • Chemical Formula: LiNO3
  • Molecular Weight: 68.95
  • Purity: 99%
  • Melting Point: 251°C

Chemical Identifiers

CAS 7790-69-4

Specifications

Melting Point 251°C
Color White
CAS Max % 1
Physical Form Crystals or Needles
Quantity 2.5 kg
Assay Percent Range 99.00%
Molecular Formula LiNO3
UN Number UN2722
Synonym Nitric Acid, Lithium Salt
Solubility Information Soluble in water
InChI Formula InChI=1S/Li.NO3/c;2-1(3)4/q+1;-1
SMILES [Li+].[O-][N+](=O)[O-]
IUPAC Name lithium nitrate
Molecular Weight (g/mol) 68.95
Formula Weight 68.95
Percent Purity 99%
Grade Reagent
Odor Odorless
Packaging Amber Glass Bottle
Sensitivity Deliquescent
Appearance Crystals, Granular
Specific Gravity 1.2 to 1.3
Decomposition Information Nitrogen Oxides (NOx), Lithium Oxides
Chemical Name or Material Lithium Nitrate, Crystal
Show More Show Less

Caution: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.