accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaOWOOO

CAS RN 7790-75-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 287.91
Molecular Formula: CaO4W
InChi Key: FDMFQOCGNBYKPY-UHFFFAOYSA-N
SMILES: [Ca++].[O-][W]([O-])(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)