Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calcium tungstate

Catalog No. 248665500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-866-5500G 500 g
24-866-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-866-5500G Supplier Sigma Aldrich Supplier No. 248665500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Calcium tungstate (CaWO4) is an optical material, which can be used as a laser host material for a variety of electronic applications. It has a scheelite structure with luminescence, and thermo-luminescence properties.CaWO4 can be used in the fabrication of a radio-sensitizer for cancer radiotherapy applications. It can also be used as a catalyst for the oxidation of organic substrates with hydrogen peroxide.

Specifications

CAS 7790-75-2
Density 6.06 g/mL (at 25°C)
Molecular Formula CaO4W
Linear Formula CaWO4
Quantity 500 g
MDL Number MFCD00010913
Molecular Weight (g/mol) 287.92
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.