CAS RN 7793-13-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cobalt (II) Chloride Hexahydrate, Reagent Grade, Innovating Science™
CAS: 7793-13-1 Molecular Weight (g/mol): 297.93 g/mol