accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 779331-47-4

IUPAC Name: [2-(benzyloxy)-5-fluorophenyl]boronic acid
Synonyms: 2-(benzyloxy)-5-fluorophenylboronic acid
Molecular Weight (g/mol): 246.04
Molecular Formula: C13H12BFO3
InChi Key: ULIJNMCAPASXHA-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=CC=C1OCC1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results