accessibility menu, dialog, popup

Test

Viewing profile as user:

CAS RN 77998-61-9

CH3SiH3CCH3SBr

CAS RN 77998-61-9

IUPAC Name: (4-bromothiophen-2-yl)trimethylsilane
Synonyms: (4-bromothiophen-2-yl)trimethylsilane
Molecular Weight (g/mol): 235.21
Molecular Formula: C7H11BrSSi
InChi Key: PHSJATTVQJWOPL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC(Br)=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results