Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-(trimethylsilyl)thiophene 97.0+%, TCI America™
SDP

Catalog No. B4856200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4856200MG 200 mg
B48561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4856200MG Supplier TCI America Supplier No. B4856200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77998-61-9
Molecular Formula C7H11BrSSi
Molecular Weight (g/mol) 235.21
MDL Number MFCD28657911
InChI Key PHSJATTVQJWOPL-UHFFFAOYSA-N
Synonym (4-Bromo-2-thienyl)trimethylsilane
PubChem CID 91972119
IUPAC Name (4-bromothiophen-2-yl)trimethylsilane
SMILES C[Si](C)(C)C1=CC(Br)=CS1

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 235.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-(trimethylsilyl)thiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.