accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78-09-1

IUPAC Name: (triethoxymethoxy)ethane
Synonyms: (triethoxymethoxy)ethane
Molecular Weight (g/mol): 192.26
Molecular Formula: C9H20O4
InChi Key: CWLNAJYDRSIKJS-UHFFFAOYSA-N
SMILES: CCOC(OCC)(OCC)OCC
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)