accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CCH3

CAS RN 78-59-1

IUPAC Name: 3,5,5-trimethylcyclohex-2-en-1-one
Synonyms: isophorone|isoforone|isoacetophorone|isoforon|diisophorone|izoforon|isooctopherone|caswell no. 506|epa pesticide chemical code 047401
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: HJOVHMDZYOCNQW-UHFFFAOYSA-N
SMILES: CC1=CC(=O)CC(C)(C)C1
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (2)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)