Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophorone, 97%

Catalog No. AAA148440F
Change view
Click to view available options
Quantity:
250 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA148440F 2500 mL
AAA14844AK 250 mL
AAA14844AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA148440F Supplier Thermo Scientific Chemicals Supplier No. A148440F
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 78-59-1 | C9H14O | 138.21 g/mol

Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings

Solubility
Soluble in water (12g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-59-1
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00001584
InChI Key HJOVHMDZYOCNQW-UHFFFAOYSA-N
Synonym isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one
PubChem CID 6544
ChEBI CHEBI:34800
IUPAC Name 3,5,5-trimethylcyclohex-2-en-1-one
SMILES CC1=CC(=O)CC(C)(C)C1

Specifications

Melting Point -8°C
Density 0.922
Boiling Point 99°C (18 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.478
Quantity 2500 mL
Beilstein 1280721
Merck Index 14,5196
Solubility Information Soluble in water (12g/L).
Formula Weight 138.21
Percent Purity 97%
Chemical Name or Material Isophorone
Show More Show Less
Hazard Category H227-H302+H312-H319-H335-H351
Hazard Statement GHS H Statement
H351-H302-H312-H319-H335-H227
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P370+P378q-P501c
EINECSNumber 201-126-0
RTECSNumber GW7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.