accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HO

CAS RN 7803-49-8

IUPAC Name: hydroxylamine
Synonyms: hydroxylamine oxammonium dihydridohydroxidonitrogen hydroxylamine (substance) H2nho (1) hydroxylamine
Molecular Weight (g/mol): 33.03
Molecular Formula: H3NO
InChi Key: AVXURJPOCDRRFD-UHFFFAOYSA-N
SMILES: NO
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)