Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydroxylamine Solution

Catalog No. 46780450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-780-450ML 50 mL
46-780-410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 46-780-450ML Supplier Sigma Aldrich Supplier No. 46780450ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Prodrug for cardiovascular agent N-d-hydroxy-L-arginine (NOHA, nitric oxide precursor)Hydroxyaminoguanidines as anti-cancer agentsNonsteroidal 2,3-dihydroquinoline glucocorticoid receptor agonists with reduced phosphoenolpyruvate caboxykinase (PEPCK) transactivationCarboxamide derivatives of ofloxacin with improved antimicrobial propertiesAnalogues of coumarin based TNF-+a converting enzyme (TACE) inhibitorsHIV integrase inhibitors

Specifications

CAS 7803-49-8
Density 1.078 g/mL at 25°C
Boiling Point 107°C
Molecular Formula H3NO
Linear Formula NH2OH
MDL Number MFCD00044522
Quantity 50 mL
Synonym HDA
Molecular Weight (g/mol) 33.03 g/mol
Biological Activity Blood, Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.