CAS RN 78033-08-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ 8-Methoxymethyl-3-isobutyl-1-methylxanthine
A cell-permeable selective inhibitor of Ca2+-calmodulin-dependent phosphodiesterase (PDE I; IC50 = 4 μM).