Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 8-Methoxymethyl-3-isobutyl-1-methylxanthine

Catalog No. 45420210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-420-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-420-210MG Supplier MilliporeSigma™ Supplier No. 45420210MG
Add to Cart
Add to Cart

A cell-permeable selective inhibitor of Ca2+-calmodulin-dependent phosphodiesterase (PDE I; IC50 = 4 μM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins PDE 1
CAS 78033-08-6
Molecular Formula C12H18N4O3
Formula Weight 266.3g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.