accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPOClOCH3

CAS RN 78055-64-8

IUPAC Name: diethyl [(3-chlorophenyl)methyl]phosphonate
Synonyms: diethyl (3-chlorophenyl)methylphosphonate
Molecular Weight (g/mol): 262.67
Molecular Formula: C11H16ClO3P
InChi Key: XJVQZLBCYMWOSM-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC(Cl)=CC=C1)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results