Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (3-Chlorobenzyl)phosphonate 98.0+%, TCI America™
SDP

Catalog No. C15955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15955G 5 g
C15951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15955G Supplier TCI America Supplier No. C15955G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78055-64-8
Molecular Formula C11H16ClO3P
Molecular Weight (g/mol) 262.67
MDL Number MFCD01861282
InChI Key XJVQZLBCYMWOSM-UHFFFAOYSA-N
Synonym (3-Chlorobenzyl)phosphonic Acid Diethyl Ester
PubChem CID 10106792
IUPAC Name 1-chloro-3-(diethoxyphosphorylmethyl)benzene
SMILES CCOP(=O)(CC1=CC(=CC=C1)Cl)OCC

Specifications

Color Colorless
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 163°C
Formula Weight 262.67
Percent Purity ≥98.0% (GC)
Chemical Name or Material Diethyl (3-Chlorobenzyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.