CAS RN 780802-33-7
IUPAC Name:
3-(piperidin-1-yl)pyridine-2-carbonitrile
Synonyms:
3-(piperidin-1-yl)pyridine-2-carbonitrile
Molecular Weight (g/mol):
187.25
Molecular Formula:
C11H13N3
InChi Key:
IUXFRKMLWJPBRT-UHFFFAOYSA-N
SMILES:
N#CC1=C(C=CC=N1)N1CCCCC1
1
–
1
of
1
results
1
3-Piperidin-1-ylpyridine-2-carbonitrile, 97%, Thermo Scientific™
CAS: 780802-33-7 Molecular Formula: C11H13N3 Molecular Weight (g/mol): 187.25 MDL Number: MFCD08690241 InChI Key: IUXFRKMLWJPBRT-UHFFFAOYSA-N Synonym: 3-piperidin-1-yl pyridine-2-carbonitrile,2-pyridinecarbonitrile,3-1-piperidinyl,3-piperidylpyridine-2-carbonitrile,2-pyridinecarbonitrile, 3-1-piperidinyl,3,4,5,6-tetrahydro-2h-1,3' bipyridinyl-2'-carbonitrile PubChem CID: 23016570 IUPAC Name: 3-piperidin-1-ylpyridine-2-carbonitrile SMILES: N#CC1=C(C=CC=N1)N1CCCCC1