accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78081-87-5

IUPAC Name: 2,6-bis({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 2,6-bis({[(9H-fluoren-9-ylmethoxy)carbonyl]amino})hexanoic acid
Molecular Weight (g/mol): 590.68
Molecular Formula: C36H34N2O6
InChi Key: BMJRTKDVFXYEFS-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)