Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-alpha,N-epsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine 98.0+%, TCI America™
SDP

Catalog No. B493125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B493125G 25 g
B49315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B493125G Supplier TCI America Supplier No. B493125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78081-87-5
Molecular Formula C36H34N2O6
Molecular Weight (g/mol) 590.676
MDL Number MFCD00080275
InChI Key BMJRTKDVFXYEFS-XIFFEERXSA-N
Synonym fmoc-lys fmoc-oh, n,n'-bis-fmoc-l-lysine, fmoc-l-lys fmoc-oh, n, a,n, a-di-fmoc-l-lysine, 2s-2,6-bis 9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoclys fmoc oh, pubchem18973, n, n'-bis-fmoc-l-lysine, n-a,n-epsilon-di-fmoc-l-lysine, nalpha,nepsilon-di-fmoc-l-lysine
PubChem CID 13783708
IUPAC Name (2S)-2,6-bis(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 135°C
Color White
Quantity 25 g
Formula Weight 590.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-alpha,N-epsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.