accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 781-35-1

IUPAC Name: 1,1-diphenylpropan-2-one
Synonyms: 2-propanone, 1,1-diphenyl-
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: DBNWBEGCONIRGQ-UHFFFAOYSA-N
SMILES: CC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results