Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diphenylacetone 99.0+%, TCI America™
SDP

Catalog No. D139725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D139725G 25 g
D139710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D139725G Supplier TCI America Supplier No. D139725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 781-35-1
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00008759
InChI Key DBNWBEGCONIRGQ-UHFFFAOYSA-N
Synonym 1,1-diphenylacetone, 1,1-diphenyl-2-propanone, 2-propanone, 1,1-diphenyl, 1,1-diphenyl acetone, methyl diphenylmethyl ketone, ketone, diphenylmethyl methyl, benzhydryl methyl ketone, diphenylacetone, 3,3-diphenyl acetone, 2-propanone, diphenyl
PubChem CID 69907
IUPAC Name 1,1-diphenylpropan-2-one
SMILES CC(=O)C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 135°C
Quantity 25 g
Formula Weight 210.28
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diphenylacetone
RTECSNumber UC2010020
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.