accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3

CAS RN 781-43-1

IUPAC Name: 9,10-dimethylanthracene
Synonyms: 9,10-dimethylanthracene
Molecular Weight (g/mol): 206.29
Molecular Formula: C16H14
InChi Key: JTGMTYWYUZDRBK-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (7)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)