Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9,10-Dimethylanthracene

Catalog No. D1467061G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1467061G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1467061G Supplier Sigma Aldrich Supplier No. D1467061G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(pyridin-2-yl)isopropyl amine (pip-amine) is a tertiary amine
  • A bidentate directing group, useful for the functionalization of c-h bond, has been developed from pip-amine.the shi auxiliary is a powerful directing group for the hydroxylation of arenes through copper-mediated c-h activation
  • This useful auxiliary can be removed through acid-mediated hydrolysis and has shown to be done on gram-scale.copper-mediated hydroxylation of arenes and heteroarenes directed by a removable bidentate auxiliary

Specifications

CAS 781-43-1
Melting Point 182°C to 184°C (lit.)
Molecular Formula C16H14
Linear Formula C16H14
MDL Number MFCD00001262
Quantity 500 mg
Molecular Weight (g/mol) 206.28
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.