accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCH3OOONOO

CAS RN 78269-85-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-(trimethylsilyl)ethyl carbonate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-(trimethylsilyl)ethyl carbonate
Molecular Weight (g/mol): 259.33
Molecular Formula: C10H17NO5Si
InChi Key: FLDNDAMSCINJDX-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCOC(=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)