Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide 98.0+%, TCI America™
SDP

Catalog No. T25915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25915G 5 g
T25911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25915G Supplier TCI America Supplier No. T25915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78269-85-9
Molecular Formula C10H17NO5Si
Molecular Weight (g/mol) 259.333
MDL Number MFCD02683467
InChI Key FLDNDAMSCINJDX-UHFFFAOYSA-N
Synonym Teoc-OSu
PubChem CID 5018386
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-trimethylsilylethyl carbonate
SMILES C[Si](C)(C)CCOC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 259.33
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.