accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78306-92-0

IUPAC Name: 2-amino-3-(naphthalen-1-yl)propanoic acid
Synonyms: alphanaphthylalanine
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: OFYAYGJCPXRNBL-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)