CAS RN 784211-09-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Cdk/Crk Inhibitor
The Cdk/Crk Inhibitor, also referenced under CAS 784211-09-2, controls the biological activity of Cdk/Crk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.