Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cdk/Crk Inhibitor

Catalog No. 2194911MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-949-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-949-11MG Supplier MilliporeSigma™ Supplier No. 2194911MG
Add to Cart
Add to Cart

The Cdk/Crk Inhibitor, also referenced under CAS 784211-09-2, controls the biological activity of Cdk/Crk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Cdk1/B, Cdk2/E, Cdk3/E, Cdk5/p35, Cdk7/H/MAT1, and Cdk9
CAS 784211-09-2
Molecular Formula C23H22Cl2N4O3
Formula Weight 473.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.