CAS RN 78491-70-0
IUPAC Name:
pentafluoroethane-1-sulfonamide
Synonyms:
pentafluoroethanesulfonamide
Molecular Weight (g/mol):
199.1
Molecular Formula:
C2H2F5NO2S
InChi Key:
QDUKVMNUAQTMQW-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Pentafluoroethanesulfonamide 98.0+%, TCI America™
CAS: 78491-70-0 Molecular Formula: C2H2F5NO2S Molecular Weight (g/mol): 199.095 MDL Number: MFCD19442187 InChI Key: QDUKVMNUAQTMQW-UHFFFAOYSA-N Synonym: Perfluoroethanesulfonamide PubChem CID: 11252905 IUPAC Name: 1,1,2,2,2-pentafluoroethanesulfonamide SMILES: C(C(F)(F)S(=O)(=O)N)(F)(F)F