accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNH2

CAS RN 78597-54-3

IUPAC Name: 5-(4-methylphenyl)-1H-pyrazol-3-amine
Synonyms: 5-(4-methylphenyl)-1H-pyrazol-3-amine
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: GVPFRVKDBZWRCZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC(N)=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results