Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-3-(4-methylphenyl)pyrazole, 97%, Thermo Scientific™

Catalog No. AC438800050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438800050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438800050 Supplier Thermo Scientific Chemicals Supplier No. 438800050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78597-54-3
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
InChI Key GVPFRVKDBZWRCZ-UHFFFAOYSA-N
Synonym 5-p-tolyl-1h-pyrazol-3-amine, 3-4-methylphenyl-1h-pyrazol-5-amine, 5-p-tolyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methylphenyl pyrazole, 3-amino-5-4-methylphenyl-1h-pyrazole, 3-amino-5-p-tolylpyrazole, 5-4-methylphenyl-1h-pyrazol-3-amine, 3-p-tolyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-4-methylphenyl, 5-4-methylphenyl-2h-pyrazol-3-amine
PubChem CID 736768
IUPAC Name 5-(4-methylphenyl)-1H-pyrazol-3-amine
SMILES CC1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.3°C to 153.3°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 173.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Amino-3-(4-methylphenyl)pyrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.