accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78667-04-6

IUPAC Name: 2-(chloromethyl)-1-methyl-1H-imidazole hydrochloride
Synonyms: 2-(chloromethyl)-1-methylimidazole hydrochloride
Molecular Weight (g/mol): 167.03
Molecular Formula: C5H8Cl2N2
InChi Key: JDIPKQBKSBWIKU-UHFFFAOYSA-N
SMILES: Cl.CN1C=CN=C1CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results