Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride, 97%, Thermo Scientific™

Catalog No. CC20874DE
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC20874DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC20874DE Supplier Thermo Scientific Chemicals Supplier No. CC20874DE

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78667-04-6
Molecular Formula C5H8Cl2N2
Molecular Weight (g/mol) 167.033
InChI Key JDIPKQBKSBWIKU-UHFFFAOYSA-N
Synonym 2-chloromethyl-1-methyl-1h-imidazole hydrochloride, 2-chloromethyl-1-methylimidazole hydrochloride, 1h-imidazole, 2-chloromethyl-1-methyl-, monohydrochloride, 2-chloromethyl-1-methyl-1h-imidazole hcl, 1-methyl-2-chloromethylimidazole hydrochloride, 2-chloromethyl-1-methylimidazole, chloride, 2-chloromethyl-1-methyl imidazole hydrochloride, 1-methyl-2-chloromethyl imidazole hydrochloride
PubChem CID 2795406
IUPAC Name 2-(chloromethyl)-1-methylimidazole;hydrochloride
SMILES CN1C=CN=C1CCl.Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 167.04
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.