accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78739-01-2

IUPAC Name: (2R)-2-azaniumyl-4-phosphonatobutanoate
Synonyms: (2R)-2-ammonio-4-phosphonatobutanoate
Molecular Weight (g/mol): 181.09
Molecular Formula: C4H8NO5P
InChi Key: DDOQBQRIEWHWBT-GSVOUGTGSA-L
SMILES: [NH3+][C@H](CCP([O-])([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results