accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3CSOOHNNNCH3OSiCH3H3CCH3H3CCH3

CAS RN 787554-04-5

IUPAC Name: N-{1-[(tert-butyldiphenylsilyl)oxy]-3-(1-methyl-1H-imidazol-5-yl)propan-2-yl}-2,4,6-tris(propan-2-yl)benzene-1-sulfonamide
Synonyms: N-{1-[(tert-butyldiphenylsilyl)oxy]-3-(3-methylimidazol-4-yl)propan-2-yl}-2,4,6-triisopropylbenzenesulfonamide
Molecular Weight (g/mol): 660.01
Molecular Formula: C38H53N3O3SSi
InChi Key: WRQMRAGSDVXWEV-UHFFFAOYNA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)NC(CO[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C(C)(C)C)CC1=CN=CN1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results