Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(2,4,6-Triisopropylbenzenesulfonyl)-O-(tert-butyldiphenylsilyl)-pros-methyl-L-histidinol 98.0+%, TCI America™
SDP

Catalog No. T2223100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2223100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2223100MG Supplier TCI America Supplier No. T2223100MG

Chemical Identifiers

CAS 787554-04-5
Molecular Formula C38H53N3O3SSi
Molecular Weight (g/mol) 660.005
InChI Key WRQMRAGSDVXWEV-HKBQPEDESA-N
PubChem CID 11193139
IUPAC Name N-[(2S)-1-[tert-butyl(diphenyl)silyl]oxy-3-(3-methylimidazol-4-yl)propan-2-yl]-2,4,6-tri(propan-2-yl)benzenesulfonamide
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)NC(CC2=CN=CN2C)CO[Si](C3=CC=CC=C3)(C4=CC=CC=C4)C(C)(C)C)C(C)C

Specifications

Melting Point 177°C
Quantity 100 mg
Formula Weight 660.01
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(2,4,6-Triisopropylbenzenesulfonyl)-O-(tert-butyldiphenylsilyl)-pros-methyl-L-histidinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.