accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 78782-17-9

IUPAC Name: 4,4,5,5-tetramethyl-2-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 267.97
Molecular Formula: C13H26B2O4
InChi Key: MQYZGGWWHUGYDR-UHFFFAOYSA-N
SMILES: CC1(C)OB(CB2OC(C)(C)C(C)(C)O2)OC1(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results