Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[(pinacolato)boryl]methane 95.0+%, TCI America™
SDP

Catalog No. B41035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41035G 5 g
B41031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41035G Supplier TCI America Supplier No. B41035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 78782-17-9
Molecular Formula C13H26B2O4
Molecular Weight (g/mol) 267.97
MDL Number MFCD27977747
InChI Key MQYZGGWWHUGYDR-UHFFFAOYSA-N
PubChem CID 11311685
IUPAC Name 4,4,5,5-tetramethyl-2-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(CB2OC(C)(C)C(C)(C)O2)OC1(C)C

Specifications

Melting Point 53°C
Color White
Quantity 5 g
Synonym 2,2′-Methylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methane
Formula Weight 267.97
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Bis[(pinacolato)boryl]methane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.