accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CMgBr

CAS RN 78887-70-4

IUPAC Name: magnesium(2+) pentadecan-1-ide bromide
Synonyms: magnesium(2+) pentadecan-1-ide bromide
Molecular Weight (g/mol): 315.62
Molecular Formula: C15H31BrMg
InChi Key: FAXYNDJJWMSBJG-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CCCCCCCCCCCCCC[CH2-]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)