Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecylmagnesium bromide, 0.4M solution in THF, AcroSeal™

Catalog No. AC436160500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436160500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436160500 Supplier Thermo Scientific Chemicals Supplier No. 436160500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 78887-70-4 | C15H31BrMg | 315.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78887-70-4 , 60-29-7
Molecular Formula C15H31BrMg
Molecular Weight (g/mol) 315.62
MDL Number MFCD00209607
InChI Key FAXYNDJJWMSBJG-UHFFFAOYSA-M
Synonym n-pentadecylmagnesium bromide, pentadecylmagnesium bromide solution, pentadecylmagnesiumbromide, magnesium,bromopentadecyl, pentadecylmagnesium bromide, nnibywyqyujvfg-uhfffaoysa-m, pentadecylmagnesium bromide solution, 0.5 m in diethyl ether, grignard reagent
PubChem CID 4641098
IUPAC Name magnesium;pentadecane;bromide
SMILES [Mg++].[Br-].CCCCCCCCCCCCCC[CH2-]

Specifications

Color Yellow to Brown
Density 0.793g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)14MgBr
Quantity 50 mL
Solubility Information Solubility in water: reacts.
Formula Weight 315.62
Physical Form Liquid
Chemical Name or Material Pentadecylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.