accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OH2C

CAS RN 79-06-1

IUPAC Name: prop-2-enamide
Synonyms: polyacrylamide polyhall 27 versicol W 11 polyacrylamide solution amide propenoic acid acrylamide (substance) flokonit e amid kyseliny akrylove sursolan P 5 polyacrylamide resin
Molecular Weight (g/mol): 71.08
Molecular Formula: C3H5NO
InChi Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES: NC(=O)C=C
Quantity 
  • (7)
  • (3)
  • (1)
  • (10)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (15)
  • (5)
Molecular Weight (g/mol) 
  • (52)
  • (1)
Physical Form 
  • (2)
  • (12)
  • (6)
  • (1)
  • (17)
  • (4)
  • (9)
Color 
  • (16)
  • (18)
Grade 
  • (10)
  • (1)
  • (14)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (26)
  • (5)
  • (1)
  • (1)
  • (7)
  • (1)
  • (8)
  • (1)
  • (5)
  • (1)
  • (5)
  • (29)
  • (3)
  • (7)
  • (3)
  • (1)
  • (10)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (15)
  • (5)
  • (52)
  • (1)
  • (2)
  • (12)
  • (6)
  • (1)
  • (17)
  • (4)
  • (9)
  • (16)
  • (18)
  • (10)
  • (1)
  • (14)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (4)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (8)
  • (3)
  • (5)
  • (1)
  • (1)
  • (4)
  • (1)