Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide, Ultra Pure, MP Biomedicals™

Catalog No. MP04814326 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04814326 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP04814326 Supplier MP Biomedicals, Inc Supplier No. 04814326
Add to Cart
Add to Cart

Acrylamide Ultra Pure, >99.9%, White Crystalline Powder

  • Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers.
  • It polymerizes in the presence of free radicals in aqueous solutions.
  • It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym 2-Propenamide, Acrylic acid amide
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Crystalline Powder
Boiling Point 125°C/25 mmHg (literature)
Melting Point 82°C to 92°C
Conductivity ≤10μmho (35% solution) (Lit.)
Color White
Quantity 500 g
Flash Point 280.4°F / 138°C (Lit.)
UN Number UN2074
Beilstein 605349
Solubility Information Soluble in water (40% aq soln-Clear, Colorless Solution), methanol (155g/100 mL), ethanol (86g/100 mL), alcohol, acetone (63.1g/100 mL); only minimally soluble in benzene or heptane.
Grade >99.9%
Show More Show Less
RTECSNumber AS3325000
storageNote1 Store at Room Temperature (15-30°C), Dessicated.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.