accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHO

CAS RN 79-14-1

IUPAC Name: 2-hydroxyacetic acid
Synonyms: glycolic acid glycocide kyselina hydroxyoctova 2-(14)c-labeled glycolic acid caswell no. 470 1-(14)c-labeled glycolic acid calcium salt glycolic acid monoammonium salt glycolic acid glycollate monopotassium salt glycolic acid
Molecular Weight (g/mol): 76.05
Molecular Formula: C2H4O3
InChi Key: AEMRFAOFKBGASW-UHFFFAOYSA-N
SMILES: OCC(O)=O
Molecular Weight (g/mol) 
  • (16)
  • (5)
  • (15)
Quantity 
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (4)
Grade 
  • (1)
  • (7)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (22)
  • (11)
  • (3)
  • (11)
  • (11)
  • (4)
  • (11)
  • (16)
  • (5)
  • (15)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (4)
  • (1)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (4)
  • (1)
  • (7)
  • (4)
  • (2)