Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycolic Acid, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019204 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9204 100 mg
11-101-7918 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9204 Supplier MilliporeSigma Supelco Supplier No. 94815100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 79-14-1
Product Line TraceCERT
Physical Form Neat
Linear Formula HOCH2COOH
MDL Number MFCD00004312
Melting Point 75°C to 80°C
Synonym Hydroxyacetic acid
Quantity 100 mg
Formula Weight 76.05
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.