CAS RN 79-29-8
2,3-Dimethylbutane 98.0+%, TCI America™
CAS: 79-29-8 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00008925 InChI Key: ZFFMLCVRJBZUDZ-UHFFFAOYSA-N Synonym: diisopropyl,biisopropyl,butane, 2,3-dimethyl,1,1,2,2-tetramethylethane,hsdb 76,unii-68isq7a432,ccris 6020,ch3 2chch ch3 2,acmc-1bmfq,dsstox_cid_5112 PubChem CID: 6589 IUPAC Name: 2,3-dimethylbutane SMILES: CC(C)C(C)C
2,3-Dimethylbutane, 98+%
CAS: 79-29-8 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00008925 InChI Key: ZFFMLCVRJBZUDZ-UHFFFAOYSA-N Synonym: diisopropyl,biisopropyl,butane, 2,3-dimethyl,1,1,2,2-tetramethylethane,hsdb 76,unii-68isq7a432,ccris 6020,ch3 2chch ch3 2,acmc-1bmfq,dsstox_cid_5112 PubChem CID: 6589 IUPAC Name: 2,3-dimethylbutane SMILES: CC(C)C(C)C
2,3-Dimethylbutane, 99%
CAS: 79-29-8 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00008925 InChI Key: ZFFMLCVRJBZUDZ-UHFFFAOYSA-N Synonym: diisopropyl,biisopropyl,butane, 2,3-dimethyl,1,1,2,2-tetramethylethane,hsdb 76,unii-68isq7a432,ccris 6020,ch3 2chch ch3 2,acmc-1bmfq,dsstox_cid_5112 PubChem CID: 6589 IUPAC Name: 2,3-dimethylbutane SMILES: CC(C)C(C)C
2,3-Dimethylbutane, 98+%
CAS: 79-29-8 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00008925 InChI Key: ZFFMLCVRJBZUDZ-UHFFFAOYSA-N Synonym: diisopropyl,biisopropyl,butane, 2,3-dimethyl,1,1,2,2-tetramethylethane,hsdb 76,unii-68isq7a432,ccris 6020,ch3 2chch ch3 2,acmc-1bmfq,dsstox_cid_5112 PubChem CID: 6589 IUPAC Name: 2,3-dimethylbutane SMILES: CC(C)C(C)C
2,3-Dimethylbutane, SPEX CertiPrep™
CAS: 79-29-8 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 InChI Key: ZFFMLCVRJBZUDZ-UHFFFAOYSA-N PubChem CID: 6589 IUPAC Name: 2,3-dimethylbutane SMILES: CC(C)C(C)C
Sigma Aldrich 2,3-Dimethylbutane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.