accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2ONaNaCH3NH3C(S)(R)(S)OH(R)COO(S)CH3HOO(S)OHOH2NOHO(Z)

CAS RN 79-57-2

IUPAC Name: disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate
Synonyms: oxytetracycline
Molecular Weight (g/mol): 540.43
Molecular Formula: C22H26N2Na2O11
InChi Key: OLRBUPVCLOPDOT-GETZKYKJSA-M
SMILES: O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Color 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)